Search
Search

P2RX1| Gene Symbol: P2RX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002558.3 | NP_002549.1 | 6 | 1030 | ||
| XM_006721529.2 | XP_006721592.1 | 6 | 819 | |||
| XM_011523896.2 | XP_011522198.1 | 7 | 1422 | |||
| XM_011523897.2 | XP_011522199.1 | 6 | 870 | |||
| XM_011523898.2 | XP_011522200.1 | 6 | 1483 | |||
| XM_011523899.2 | XP_011522201.1 | 6 | 1534 | |||
| XM_011523900.2 | XP_011522202.1 | 6 | 1706 | |||
| GenBank mRNA | AF020498.1 | - | 6 | 865 | ||
| AK313264.1 | - | 6 | 829 | |||
| BC027949.1 | - | 5 | 1112 | |||
| BC044657.1 | - | 6 | 805 | 5756349 | ||
| U45448.1 | - | 6 | 785 | |||
| X83688.1 | - | 6 | 762 | |||