Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CTSB SIRNA Assays ›
          Gene Symbol:
          CTSB
          Gene Name:
          cathepsin B
          Gene Aliases:
          APPS, CPSB
          Chromosome Location:
          Chr.8: 11842524 - 11868150 on Build GRCh38
          UniGene ID:
          Hs.520898
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 105578

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CTSB
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001317237.1 NP_001304166.1 3 358
          NM_001908.4 NP_001899.1 2 270
          NM_147780.3 NP_680090.1 4 432
          NM_147781.3 NP_680091.1 3 389
          NM_147782.3 NP_680092.1 3 358
          NM_147783.3 NP_680093.1 3 344
          XM_006716244.2 XP_006716307.1 4 456
          XM_006716245.2 XP_006716308.1 2 560
          XM_011543812.2 XP_011542114.1 4 365
          XM_017013097.1 XP_016868586.1 3 346
          XM_017013098.1 XP_016868587.1 3 346
          XM_017013099.1 XP_016868588.1 4 388
          XM_017013100.1 XP_016868589.1 3 290
          GenBank mRNA AK075393.1 - 2 246
          AK092070.1 - 3 281
          AK130184.1 - 2 207
          AK290239.1 - 2 207
          AK296845.1 - 2 209
          AL543654.3 - 3 318
          BC010240.1 - 3 287 4154307
          BC095408.1 - 2 207 30334082
          BG770805.1 - 4 341
          BM008741.1 - 2 246
          BX647765.1 - 3 249
          BX647952.1 - 2 210
          BX648239.1 - 1 1041
          CB997355.1 - 2 284
          DC386022.1 - 2 270
          L16510.1 - 3 272
          M14221.1 - 3 289

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          cysteine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          toll-like receptor signaling pathway
          proteolysis
          autophagy
          spermatogenesis
          skeletal muscle tissue development
          response to wounding
          response to glucose
          response to organic cyclic compound
          response to amine
          collagen catabolic process
          epithelial cell differentiation
          regulation of apoptotic process
          response to peptide hormone
          response to ethanol
          decidualization
          viral entry into host cell
          regulation of catalytic activity
          proteolysis involved in cellular protein catabolic process
          negative regulation of cell death
          response to interleukin-4
          cellular response to mechanical stimulus
          cellular response to thyroid hormone stimulus
          cysteine-type endopeptidase activity
          serine-type endopeptidase activity
          protein binding
          collagen binding
          peptidase activity
          cysteine-type peptidase activity
          kininogen binding
          peptide binding
          proteoglycan binding
          protein self-association

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-7gdsv:80/100.66.79.88:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0