Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002795.3 | NP_002786.2 | 3 | 365 | ||
NR_104194.1 | 3 | 365 | ||||
NR_104195.1 | 3 | 365 | ||||
GenBank mRNA | BC013008.2 | - | 3 | 328 | 3010175 | |
BQ428285.1 | - | 3 | 325 | |||
CN412852.1 | - | 3 | 269 | |||
D26598.1 | - | 3 | 268 |