Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other UCHL5 SIRNA Assays ›
Gene Symbol:
UCHL5
Gene Name:
ubiquitin C-terminal hydrolase L5
Gene Aliases:
CGI-70, INO80R, UCH-L5, UCH37
Chromosome Location:
Chr.1: 193012366 - 193060140 on Build GRCh38
UniGene ID:
Hs.145469
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
105723
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: UCHL5
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001199261.1
NP_001186190.1
2
251
NM_001199262.1
NP_001186191.1
2
251
NM_001199263.1
NP_001186192.1
2
251
NM_015984.3
NP_057068.1
2
251
NR_037607.1
2
517
XM_005245244.4
XP_005245301.1
2
334
XM_005245246.4
XP_005245303.1
2
334
XM_005245247.4
XP_005245304.1
2
186
XM_005245248.4
XP_005245305.1
2
209
XM_005245249.4
XP_005245306.1
2
105
XM_006711366.3
XP_006711429.1
2
334
XM_006711367.3
XP_006711430.1
2
334
XM_006711368.3
XP_006711431.1
2
334
XM_006711369.2
XP_006711432.1
2
334
XM_006711370.3
XP_006711433.3
2
287
XM_006711371.3
XP_006711434.1
2
105
XM_011509604.1
XP_011507906.1
2
334
XM_011509607.2
XP_011507909.1
2
145
XM_011509608.2
XP_011507910.1
2
225
XM_011509609.2
XP_011507911.1
2
129
XM_017001429.1
XP_016856918.1
2
334
XM_017001430.1
XP_016856919.1
2
334
XM_017001431.1
XP_016856920.1
2
334
XM_017001432.1
XP_016856921.1
2
334
XM_017001433.1
XP_016856922.1
2
334
XM_017001434.1
XP_016856923.1
2
287
XM_017001435.1
XP_016856924.1
2
287
XM_017001436.1
XP_016856925.1
2
225
XM_017001437.1
XP_016856926.1
2
225
XM_017001438.1
XP_016856927.1
2
129
XM_017001439.1
XP_016856928.1
2
550
XM_017001440.1
XP_016856929.1
2
514
XM_017001441.1
XP_016856930.1
2
490
XM_017001442.1
XP_016856931.1
2
105
XM_017001443.1
XP_016856932.1
2
105
XM_017001444.1
XP_016856933.1
2
499
GenBank mRNA
AF147717.1
-
2
249
AF151828.1
-
2
131
AF157320.1
-
2
266
AK024751.1
-
2
197
AK225794.1
-
2
184
AK296487.1
-
2
224
AK301381.1
-
2
517
AK314445.1
-
2
195
AK316064.1
-
2
195
BC015521.1
-
2
184
3912584
BC025369.2
-
2
229
3937407
BT006790.1
-
2
118
DA255130.1
-
2
251
Back To Top
More Information
Panther Classification:
Molecular Function -
cysteine protease
Gene Ontology Categories:
Function(s)
Process(es)
DNA repair
DNA recombination
transcription, DNA-templated
regulation of transcription, DNA-templated
ubiquitin-dependent protein catabolic process
negative regulation of endopeptidase activity
protein deubiquitination
lateral ventricle development
midbrain development
forebrain morphogenesis
regulation of proteasomal protein catabolic process
thiol-dependent ubiquitin-specific protease activity
endopeptidase inhibitor activity
protein binding
poly(A) RNA binding
proteasome binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0