Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301175.1 | NP_001288104.1 | 5, 6 | 674 | ||
NM_032663.4 | NP_116052.2 | 2, 3 | 300 | |||
XM_005253962.3 | XP_005254019.1 | 2, 3 | 286 | |||
XM_006719653.3 | XP_006719716.1 | 3, 4 | 373 | |||
XM_017020048.1 | XP_016875537.1 | 2, 3 | 295 | |||
XM_017020049.1 | XP_016875538.1 | 2, 3 | 286 | |||
XM_017020050.1 | XP_016875539.1 | 2, 3 | 395 | |||
XM_017020051.1 | XP_016875540.1 | 3, 4 | 251 | |||
XM_017020052.1 | XP_016875541.1 | 2, 3 | 297 | |||
GenBank mRNA | AJ586136.1 | - | 2, 3 | 191 | ||
AK027820.1 | - | 2, 3 | 265 | |||
AK097830.1 | - | 5, 6 | 674 | |||
BC004868.2 | - | 2, 3 | 290 | 3833242 |