Search
Search

KLK8| Gene Symbol: KLK8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001281431.1 | NP_001268360.1 | 5 | 822 | ||
| NM_007196.3 | NP_009127.1 | 6 | 982 | |||
| NM_144505.2 | NP_653088.1 | 6 | 1117 | |||
| NM_144506.2 | NP_653089.1 | 4 | 559 | |||
| NM_144507.2 | NP_653090.1 | 3 | 425 | |||
| NR_104008.1 | 5 | 719 | ||||
| GenBank mRNA | AB008390.2 | - | 6 | 857 | ||
| AB008927.1 | - | 5 | 959 | |||
| AB009849.1 | - | 6 | 837 | |||
| AF095742.1 | - | 6 | 982 | |||
| AY359036.1 | - | 6 | 968 | |||
| BC040887.1 | - | 7 | 1293 | 5742016 | ||
| BM845575.1 | - | 4 | 525 | |||