Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FAS SIRNA Assays ›
          Gene Symbol:
          FAS
          Gene Name:
          Fas cell surface death receptor
          Gene Aliases:
          ALPS1A, APO-1, APT1, CD95, FAS1, FASTM, TNFRSF6
          Chromosome Location:
          Chr.10: 88968429 - 89017061 on Build GRCh38
          UniGene ID:
          Hs.244139
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 105934

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: FAS
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000043.5 NP_000034.1 9 1757
          NM_001320619.1 NP_001307548.1 8 1674
          NM_152871.3 NP_690610.1 8 1694
          NM_152872.3 NP_690611.1 8 1732
          NR_028033.3 7 1585
          NR_028034.3 6 1447
          NR_028035.3 7 1510
          NR_028036.3 8 1648
          NR_135313.1 7 1565
          NR_135314.1 9 1748
          NR_135315.1 7 1501
          XM_006717819.3 XP_006717882.1 10 6439
          XM_011539764.2 XP_011538066.1 10 1828
          XM_011539765.2 XP_011538067.1 9 1553
          XM_011539766.2 XP_011538068.1 10 1568
          XM_011539767.2 XP_011538069.1 9 1491
          GenBank mRNA AB209361.1 - 10 5074
          BC012479.1 - 9 1749 4514272
          M67454.1 - 9 1633
          X63717.1 - 9 1659

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          ovarian follicle atresia
          immunoglobulin production
          renal system process
          protein complex assembly
          apoptotic process
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          activation-induced cell death of T cells
          inflammatory cell apoptotic process
          inflammatory response
          immune response
          signal transduction
          spermatogenesis
          brain development
          aging
          circadian rhythm
          extrinsic apoptotic signaling pathway via death domain receptors
          response to toxic substance
          gene expression
          regulation of gene expression
          B cell mediated immunity
          telencephalon development
          dendrite regeneration
          positive regulation of protein homooligomerization
          response to lipopolysaccharide
          tumor necrosis factor-mediated signaling pathway
          regulation of cell proliferation
          response to drug
          ovulation cycle
          regulation of apoptotic process
          chordate embryonic development
          positive regulation of apoptotic process
          negative regulation of apoptotic process
          regulation of cysteine-type endopeptidase activity involved in apoptotic process
          positive regulation of MAPK cascade
          response to peptide hormone
          negative thymic T cell selection
          regulation of lymphocyte differentiation
          regulation of myeloid cell differentiation
          response to cycloheximide
          spleen development
          negative regulation of B cell activation
          protein homooligomerization
          response to glucocorticoid
          maternal process involved in female pregnancy
          positive regulation of lymphocyte apoptotic process
          cellular response to hydrogen peroxide
          response to growth factor
          cellular response to phenylalanine
          cellular response to mechanical stimulus
          cellular response to cobalt ion
          cellular response to lithium ion
          cellular response to glucose stimulus
          cellular response to interleukin-1
          cellular response to estrogen stimulus
          cellular response to hyperoxia
          cellular response to hypoxia
          cellular response to hydrostatic pressure
          motor neuron apoptotic process
          apoptotic signaling pathway
          extrinsic apoptotic signaling pathway
          extrinsic apoptotic signaling pathway in absence of ligand
          hepatocyte apoptotic process
          activation of cysteine-type endopeptidase activity involved in apoptotic signaling pathway
          liver regeneration
          necroptotic signaling pathway
          regulation of extrinsic apoptotic signaling pathway via death domain receptors
          negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
          response to fluoride
          positive regulation of extrinsic apoptotic signaling pathway in absence of ligand
          protease binding
          signal transducer activity
          receptor activity
          tumor necrosis factor-activated receptor activity
          protein binding
          kinase binding
          protein complex binding
          identical protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0