siRNA Details
Gene Symbol: CHM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000390.3 | NP_000381.1 | 5 | 467 | ||
NM_001320959.1 | NP_001307888.1 | 5 | 464 | |||
XM_017029242.1 | XP_016884731.1 | 5 | 463 | |||
XM_017029243.1 | XP_016884732.1 | 5 | 554 | |||
XM_017029244.1 | XP_016884733.1 | 5 | 452 | |||
XM_017029245.1 | XP_016884734.1 | 5 | 554 | |||
XM_017029246.1 | XP_016884735.1 | 5 | 433 | |||
GenBank mRNA | AK291160.1 | - | 5 | 456 | ||
AK299323.1 | - | 2 | 172 | |||
AK312443.1 | - | 5 | 456 | |||
DA403806.1 | - | 5 | 464 | |||
DB231066.1 | - | 5 | 467 | |||
X78121.1 | - | 5 | 464 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
