siRNA Details
Gene Symbol: APP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000484.3 | NP_000475.1 | 12, 13 | 1795 | ||
NM_001136016.3 | NP_001129488.1 | 11, 12 | 1719 | |||
NM_001136129.2 | NP_001129601.1 | 9, 10 | 1402 | |||
NM_001136130.2 | NP_001129602.1 | 11, 12 | 1627 | |||
NM_001136131.2 | NP_001129603.1 | 10, 11 | 1457 | |||
NM_001204301.1 | NP_001191230.1 | 12, 13 | 1795 | |||
NM_001204302.1 | NP_001191231.1 | 11, 12 | 1738 | |||
NM_001204303.1 | NP_001191232.1 | 10, 11 | 1570 | |||
NM_201413.2 | NP_958816.1 | 11, 12 | 1738 | |||
NM_201414.2 | NP_958817.1 | 10, 11 | 1570 | |||
GenBank mRNA | A02759.1 | - | 10, 11 | 1516 | ||
AF282245.1 | - | 9, 10 | 1329 | |||
AK294534.1 | - | 11, 12 | 1574 | |||
AK295373.1 | - | 2, 3 | 295 | |||
AK295621.1 | - | 11, 12 | 1680 | |||
AK296229.1 | - | 10, 11 | 1457 | |||
AK297229.1 | - | 8, 9 | 933 | |||
AK297412.1 | - | 9, 10 | 1299 | |||
AK298861.1 | - | 9, 10 | 1472 | |||
AK312326.1 | - | 12, 13 | 1660 | |||
BC065529.1 | - | 11, 12 | 1669 | 6152423 | ||
M16765.1 | - | 5, 6 | 530 | |||
M18734.1 | - | 1, 2 | 78 | |||
M28373.1 | - | 9, 10 | 1301 | |||
X06989.1 | - | 11, 12 | 1662 | |||
Y00264.1 | - | 10, 11 | 1517 |

More Information
Other Species Targeted

Panther Classification:
