Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other APP SIRNA Assays ›
          Gene Symbol:
          APP
          Gene Name:
          amyloid beta precursor protein
          Gene Aliases:
          AAA, ABETA, ABPP, AD1, APPI, CTFgamma, CVAP, PN-II, PN2
          Chromosome Location:
          Chr.21: 25880550 - 26171128 on Build GRCh38
          UniGene ID:
          Hs.434980
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 106122

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: APP
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000484.3 NP_000475.1 18 3410
          NM_001136016.3 NP_001129488.1 17 3334
          NM_001136129.2 NP_001129601.1 15 3017
          NM_001136130.2 NP_001129602.1 17 3242
          NM_001136131.2 NP_001129603.1 16 3072
          NM_001204301.1 NP_001191230.1 17 3356
          NM_001204302.1 NP_001191231.1 16 3299
          NM_001204303.1 NP_001191232.1 15 3131
          NM_201413.2 NP_958816.1 17 3353
          NM_201414.2 NP_958817.1 16 3185
          GenBank mRNA A02759.1 - 16 3131
          BC065529.1 - 17 3284 6152423
          BE907745.1 - 1 188
          S41243.1 - 1 115
          Y00264.1 - 16 3132

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          protease inhibitor

          Gene Ontology Categories:

          Function(s) Process(es)

          response to yeast
          suckling behavior
          platelet degranulation
          mRNA polyadenylation
          regulation of translation
          protein phosphorylation
          cellular copper ion homeostasis
          endocytosis
          response to oxidative stress
          cell adhesion
          regulation of epidermal growth factor-activated receptor activity
          Notch signaling pathway
          axonogenesis
          mating behavior
          locomotory behavior
          axo-dendritic transport
          cholesterol metabolic process
          adult locomotory behavior
          visual learning
          cellular process
          response to lead ion
          negative regulation of endopeptidase activity
          positive regulation of peptidase activity
          positive regulation of G2/M transition of mitotic cell cycle
          axon midline choice point recognition
          neuron remodeling
          dendrite development
          antibacterial humoral response
          antifungal humoral response
          extracellular matrix organization
          forebrain development
          neuron projection development
          ionotropic glutamate receptor signaling pathway
          regulation of multicellular organism growth
          regulation of protein binding
          cellular protein metabolic process
          innate immune response
          negative regulation of neuron differentiation
          positive regulation of mitotic cell cycle
          positive regulation of transcription from RNA polymerase II promoter
          collateral sprouting in absence of injury
          regulation of synapse structure or activity
          defense response to Gram-negative bacterium
          defense response to Gram-positive bacterium
          neuromuscular process controlling balance
          synaptic growth at neuromuscular junction
          neuron apoptotic process
          smooth endoplasmic reticulum calcium ion homeostasis
          cellular response to cAMP
          cellular response to norepinephrine stimulus
          amyloid fibril formation
          cellular response to nerve growth factor stimulus
          DNA binding
          serine-type endopeptidase inhibitor activity
          receptor binding
          protein binding
          heparin binding
          peptidase activator activity
          enzyme binding
          identical protein binding
          transition metal ion binding
          PTB domain binding
          growth factor receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0