Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LRP6 SIRNA Assays ›
          Gene Symbol:
          LRP6
          Gene Name:
          LDL receptor related protein 6
          Gene Aliases:
          ADCAD2, STHAG7
          Chromosome Location:
          Chr.12: 12116025 - 12267021 on Build GRCh38
          UniGene ID:
          Hs.584775
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 106767

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: LRP6
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_002336.2 NP_002327.2 4, 5 983
          XM_006719078.3 XP_006719141.1 4, 5 1127
          XM_011520671.2 XP_011518973.1 3, 4 733
          XM_017019304.1 XP_016874793.1 2, 3 801
          GenBank mRNA AB209683.1 - 3, 4 435
          AF074264.1 - 4, 5 918
          BC117136.1 - 4, 5 875 40125687
          BC126405.1 - 4, 5 875 8992121
          BC143725.1 - 4, 5 875 9052239

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          apolipoprotein

          Gene Ontology Categories:

          Function(s) Process(es)

          neural tube closure
          negative regulation of protein phosphorylation
          pericardium morphogenesis
          negative regulation of protein kinase activity
          positive regulation of cytosolic calcium ion concentration
          chemical synaptic transmission
          embryonic pattern specification
          neural crest formation
          neural crest cell differentiation
          Wnt signaling pathway
          single organismal cell-cell adhesion
          cerebellum morphogenesis
          thalamus development
          cerebral cortex development
          midbrain-hindbrain boundary development
          negative regulation of smooth muscle cell apoptotic process
          external genitalia morphogenesis
          odontogenesis of dentin-containing tooth
          response to peptide hormone
          Wnt signaling pathway involved in dorsal/ventral axis specification
          canonical Wnt signaling pathway involved in neural crest cell differentiation
          canonical Wnt signaling pathway involved in regulation of cell proliferation
          positive regulation of cell cycle
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of sequence-specific DNA binding transcription factor activity
          palate development
          convergent extension
          embryonic retina morphogenesis in camera-type eye
          canonical Wnt signaling pathway
          face morphogenesis
          trachea cartilage morphogenesis
          cellular response to cholesterol
          dopaminergic neuron differentiation
          negative regulation of protein serine/threonine kinase activity
          protein localization to plasma membrane
          primitive streak formation
          negative regulation of canonical Wnt signaling pathway
          receptor-mediated endocytosis of low-density lipoprotein particle involved in cholesterol transport
          Wnt signaling pathway involved in somitogenesis
          axis elongation involved in somitogenesis
          positive regulation of canonical Wnt signaling pathway
          toxin transport
          beta-catenin destruction complex disassembly
          midbrain dopaminergic neuron differentiation
          Wnt signaling pathway involved in midbrain dopaminergic neuron differentiation
          positive regulation of Wnt signaling pathway involved in dorsal/ventral axis specification
          low-density lipoprotein receptor activity
          receptor binding
          frizzled binding
          protein binding
          Wnt-protein binding
          kinase inhibitor activity
          toxin transporter activity
          apolipoprotein binding
          identical protein binding
          protein homodimerization activity
          Wnt-activated receptor activity
          coreceptor activity involved in Wnt signaling pathway
          coreceptor activity involved in canonical Wnt signaling pathway

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0