Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
Unity Lab Services
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NGFR SIRNA Assays ›
Gene Symbol:
NGFR
Gene Name:
nerve growth factor receptor
Gene Aliases:
CD271, Gp80-LNGFR, TNFRSF16, p75(NTR), p75NTR
Chromosome Location:
Chr.17: 49495293 - 49515020 on Build GRCh38
UniGene ID:
Hs.415768
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
106842
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NGFR
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_002507.3
NP_002498.1
6
2660
GenBank mRNA
BC050309.1
-
6
2660
5263715
BQ639556.1
-
1
48
M14764.1
-
6
2648
Back To Top
More Information
Panther Classification:
Molecular Function -
transmembrane signal receptor
Gene Ontology Categories:
Function(s)
Process(es)
intracellular protein transport
activation of cysteine-type endopeptidase activity involved in apoptotic process
inflammatory response
immune response
axon guidance
response to wounding
negative regulation of neuron projection development
sensory perception of pain
membrane protein intracellular domain proteolysis
positive regulation of myelination
positive regulation of synaptic transmission, cholinergic
response to lipopolysaccharide
circadian regulation of gene expression
tumor necrosis factor-mediated signaling pathway
cellular response to oxidative stress
positive regulation of Rho protein signal transduction
skeletal muscle cell differentiation
regulation of cell proliferation
glucose homeostasis
regulation of apoptotic process
positive regulation of apoptotic process
negative regulation of apoptotic process
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
regulation of cysteine-type endopeptidase activity involved in apoptotic process
positive regulation of MAPK cascade
negative regulation of neuron apoptotic process
neurotrophin TRK receptor signaling pathway
negative regulation of axonogenesis
positive regulation of axonogenesis
positive regulation of protein kinase B signaling
negative regulation of mitochondrial depolarization
positive regulation of synaptic transmission, glutamatergic
apoptotic signaling pathway
positive regulation of neuron death
regulation of reactive oxygen species metabolic process
regulation of glucose import in response to insulin stimulus
beta-amyloid binding
signal transducer activity
receptor activity
transmembrane signaling receptor activity
tumor necrosis factor-activated receptor activity
neurotrophin TRKA receptor binding
protein binding
Rab GTPase binding
ubiquitin protein ligase binding
nerve growth factor binding
preprotein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-rpnmj:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0