Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC1A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193493.1 | NP_001180422.1 | 7 | 1305 | ||
NM_003038.4 | NP_003029.2 | 8 | 2173 | |||
XM_006712079.2 | XP_006712142.1 | 8 | 1539 | |||
GenBank mRNA | AK126636.1 | - | 7 | 3363 | ||
AK315986.1 | - | 9 | 2095 | |||
BC026216.1 | - | 8 | 2074 | 4420405 | ||
BC072423.1 | - | 8 | 2133 | 6186177 | ||
L14595.1 | - | 8 | 2072 |