Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Food and Beverage
Lab Solutions
Pharma and Biopharma
Real-Time PCR
Semiconductor Analysis
Clinical and Diagnostics
Digital Solutions
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
See all services
Help and Support
Order Help
Digital Solutions
Product Support
Technical Information
Training and Education
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SOX4 SIRNA Assays ›
Gene Symbol:
SOX4
Gene Name:
SRY-box 4
Gene Aliases:
EVI16
Chromosome Location:
Chr.6: 21593741 - 21598619 on Build GRCh38
UniGene ID:
Hs.643910
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
106975
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SOX4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_003107.2
NP_003098.1
1
4111
GenBank mRNA
AJ420500.1
-
1
430
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001271205.
This siRNA also targets the following transcripts in Mouse: AK028989, AK051540, AK153771, AK031394, AK054386.
Panther Classification:
Molecular Function -
HMG box transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
skeletal system development
neural tube formation
pro-B cell differentiation
mitral valve morphogenesis
cardiac ventricle formation
cardiac right ventricle morphogenesis
atrial septum primum morphogenesis
noradrenergic neuron differentiation
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
heart development
positive regulation of cell proliferation
negative regulation of cell proliferation
glial cell proliferation
spinal cord development
spinal cord motor neuron differentiation
glial cell development
positive regulation of Wnt signaling pathway
T cell differentiation
endocrine pancreas development
negative regulation of protein ubiquitination
regulation of protein stability
positive regulation of insulin secretion
somatic stem cell population maintenance
ascending aorta morphogenesis
glucose homeostasis
DNA damage response, detection of DNA damage
positive regulation of apoptotic process
positive regulation of translation
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
negative regulation of protein export from nucleus
sympathetic nervous system development
protein stabilization
canonical Wnt signaling pathway
limb bud formation
ventricular septum morphogenesis
negative regulation of cell death
neuroepithelial cell differentiation
kidney morphogenesis
cellular response to glucose stimulus
positive regulation of canonical Wnt signaling pathway
positive regulation of N-terminal peptidyl-lysine acetylation
transcription regulatory region sequence-specific DNA binding
core promoter sequence-specific DNA binding
nucleic acid binding transcription factor activity
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
RNA polymerase II transcription coactivator activity
transcription factor activity, sequence-specific DNA binding
protein binding
protein heterodimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-72rkq:80/100.66.79.224:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline