Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SOX11 SIRNA Assays ›
Gene Symbol:
SOX11
Gene Name:
SRY-box 11
Gene Aliases:
MRD27
Chromosome Location:
Chr.2: 5692667 - 5701385 on Build GRCh38
UniGene ID:
Hs.432638
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
106976
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SOX11
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_003108.3
NP_003099.1
1
1684
GenBank mRNA
AB028641.1
-
1
1687
U23752.1
-
1
1707
Back To Top
More Information
Panther Classification:
Molecular Function -
HMG box transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
skeletal system development
kidney development
neural tube formation
lens morphogenesis in camera-type eye
outflow tract morphogenesis
cardiac ventricle formation
noradrenergic neuron differentiation
transcription from RNA polymerase II promoter
translation
positive regulation of cell proliferation
positive regulation of gene expression
negative regulation of gene expression
oligodendrocyte development
glial cell proliferation
neural crest cell development
regulation of transforming growth factor beta receptor signaling pathway
spinal cord development
glial cell development
neuron differentiation
positive regulation of BMP signaling pathway
positive regulation of hippo signaling
skeletal muscle cell differentiation
positive regulation of neuron differentiation
positive regulation of osteoblast differentiation
positive regulation of ossification
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of hormone secretion
sympathetic nervous system development
embryonic digestive tract morphogenesis
embryonic skeletal system morphogenesis
negative regulation of lymphocyte proliferation
positive regulation of neurogenesis
hard palate development
soft palate development
limb bud formation
negative regulation of glial cell proliferation
ventricular septum morphogenesis
lung morphogenesis
negative regulation of cell death
neuroepithelial cell differentiation
eyelid development in camera-type eye
somite development
cornea development in camera-type eye
closure of optic fissure
signal transduction involved in cell cycle checkpoint
positive regulation of stem cell proliferation
negative regulation of transcription regulatory region DNA binding
positive regulation of lens epithelial cell proliferation
transcription regulatory region sequence-specific DNA binding
RNA polymerase II core promoter sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
RNA polymerase II transcription coactivator activity
enhancer sequence-specific DNA binding
transcription factor activity, sequence-specific DNA binding
translation factor activity, RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0