Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TXNRD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001093771.2 | NP_001087240.1 | 8, 9 | 898 | ||
NM_001261445.1 | NP_001248374.1 | 6, 7 | 1320 | |||
NM_001261446.1 | NP_001248375.1 | 5, 6 | 875 | |||
NM_003330.3 | NP_003321.3 | 6, 7 | 1235 | |||
NM_182729.2 | NP_877393.1 | 6, 7 | 998 | |||
NM_182742.2 | NP_877419.1 | 5, 6 | 1125 | |||
NM_182743.2 | NP_877420.1 | 5, 6 | 888 | |||
GenBank mRNA | AF077367.1 | - | 5, 6 | 552 | ||
AF208018.1 | - | 4, 5 | 448 | |||
AJ001050.1 | - | 4, 5 | 461 | |||
AK128515.1 | - | 5, 6 | 783 | |||
AK226071.1 | - | 6, 7 | 706 | |||
AK293322.1 | - | 5, 6 | 535 | |||
AK295067.1 | - | 6, 7 | 634 | |||
AK296495.1 | - | 5, 6 | 525 | |||
AK304241.1 | - | 8, 9 | 881 | |||
AK308881.1 | - | 6, 7 | 929 | |||
AK312262.1 | - | 6, 7 | 897 | |||
AY057105.1 | - | 8, 9 | 874 | |||
BC018122.1 | - | 6, 7 | 611 | 3883490 | ||
BT019640.1 | - | 4, 5 | 424 | |||
CR536506.1 | - | 4, 5 | 424 | |||
D88687.1 | - | 6, 7 | 676 | |||
S79851.1 | - | 6, 7 | 863 | |||
X91247.1 | - | 6, 7 | 863 |