Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NCOA1 SIRNA Assays ›
          Gene Symbol:
          NCOA1
          Gene Name:
          nuclear receptor coactivator 1
          Gene Aliases:
          F-SRC-1, KAT13A, RIP160, SRC1, bHLHe42, bHLHe74
          Chromosome Location:
          Chr.2: 24492050 - 24770702 on Build GRCh38
          UniGene ID:
          Hs.596314
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 107233

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NCOA1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_003743.4 NP_003734.3 19 4194
          NM_147223.2 NP_671756.1 19 4194
          NM_147233.2 NP_671766.1 19 4194
          XM_005264625.1 XP_005264682.1 21 4470
          XM_005264626.1 XP_005264683.1 20 4224
          XM_005264628.1 XP_005264685.1 20 4224
          XM_017005168.1 XP_016860657.1 21 4470
          XM_017005169.1 XP_016860658.1 21 4470
          GenBank mRNA AJ000881.1 - 19 4137
          AJ000882.1 - 19 4137
          AK226063.1 - 19 4149
          AK290019.1 - 20 4224
          BC111533.1 - 18 4031 40027438
          BC111534.1 - 18 4031 40027439
          BF975824.1 - 3 468
          U19179.1 - 7 1346
          U40396.1 - 9 2852
          U59302.1 - 19 4130
          U90661.1 - 17 3978

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          histone modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of transcription from RNA polymerase II promoter by galactose
          regulation of thyroid hormone mediated signaling pathway
          transcription, DNA-templated
          lactation
          male gonad development
          bile acid and bile salt transport
          cerebellum development
          hippocampus development
          hypothalamus development
          cerebral cortex development
          androgen receptor signaling pathway
          intracellular receptor signaling pathway
          response to estradiol
          response to retinoic acid
          response to progesterone
          cellular response to hormone stimulus
          positive regulation of apoptotic process
          histone H4 acetylation
          cellular lipid metabolic process
          estrous cycle
          positive regulation of neuron differentiation
          positive regulation of transcription, DNA-templated
          positive regulation of female receptivity
          positive regulation of transcription from RNA polymerase II promoter
          male mating behavior
          labyrinthine layer morphogenesis
          cellular response to Thyroglobulin triiodothyronine
          regulation of cellular response to drug
          RNA polymerase II regulatory region DNA binding
          RNA polymerase II transcription coactivator activity
          chromatin binding
          transcription coactivator activity
          histone acetyltransferase activity
          protein binding
          ligand-dependent nuclear receptor binding
          enzyme binding
          estrogen receptor binding
          ligand-dependent nuclear receptor transcription coactivator activity
          protein complex binding
          progesterone receptor binding
          nuclear hormone receptor binding
          retinoid X receptor binding
          protein dimerization activity
          protein N-terminus binding
          androgen receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0