Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture and Transfection
Chemicals
Chromatography
Electron Microscopes
Lab Plasticware and Supplies
Lab Centrifuges
Lab Solutions
Mass Spectrometers
Next Generation Sequencers
See all product categories
Applications
Brands
Industrial and Applied Sciences
Food and Beverage
Forensics
Lab Solutions
Life Sciences
Pharma and Biopharma
Biotechnology
Clinical and Diagnostics
Digital Solutions
See all applications
Services
Lab Instrument and Equipment Services
Custom Services
Training Services
Financial and Leasing Services
Enterprise Level Lab Informatics
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Cell Biology Services
Food Safety Inspection Services
See all services
Help and Support
Contact Us
Product Documentation
Knowledge Base and Product FAQs
Learning Centers
Supply Center
eProcurement Solutions
Lab Instrument and Equipment Support
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NRP1 SIRNA Assays ›
Gene Symbol:
NRP1
Gene Name:
neuropilin 1
Gene Aliases:
BDCA4, CD304, NP1, NRP, VEGF165R
Chromosome Location:
Chr.10: 33177491 - 33334905 on Build GRCh38
UniGene ID:
Hs.131704
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
107268
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NRP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001024628.2
NP_001019799.1
6
1381
NM_001024629.2
NP_001019800.1
6
1381
NM_001244972.1
NP_001231901.1
6
1381
NM_001244973.1
NP_001231902.1
6
1381
NM_003873.5
NP_003864.4
6
1381
NR_045259.1
5
1199
XM_006717521.1
XP_006717584.1
6
1155
XM_006717522.1
XP_006717585.1
6
1155
XM_006717523.2
XP_006717586.1
6
1155
XM_006717524.1
XP_006717587.1
6
1155
XM_006717525.1
XP_006717588.1
6
1155
XM_006717526.1
XP_006717589.1
6
1155
XM_011519755.2
XP_011518057.1
6
1155
XM_011519756.1
XP_011518058.1
6
1155
XM_017016865.1
XP_016872354.1
6
1155
XM_017016866.1
XP_016872355.1
5
975
GenBank mRNA
AB209641.1
-
6
1009
AF016050.1
-
6
858
AF018956.1
-
6
858
AF145712.1
-
6
858
AF268691.1
-
6
998
AF280547.1
-
6
858
AK292822.1
-
6
1047
AL833608.1
-
5
930
AY249243.1
-
6
858
BC007533.1
-
6
1118
2958475
BC007737.2
-
6
998
4122289
BT006995.1
-
6
858
BX510902.1
-
6
1045
BX648025.1
-
7
1405
BX648250.1
-
6
1350
CR749333.1
-
6
1382
Back To Top
More Information
Panther Classification:
Molecular Function -
oxidoreductase
Gene Ontology Categories:
Function(s)
Process(es)
angiogenesis
patterning of blood vessels
neuron migration
positive regulation of endothelial cell proliferation
sprouting angiogenesis
cell migration involved in sprouting angiogenesis
signal transduction
cell-cell signaling
axon guidance
axonal fasciculation
response to wounding
organ morphogenesis
positive regulation of endothelial cell migration
positive regulation of smooth muscle cell migration
dendrite development
facial nerve structural organization
trigeminal nerve structural organization
vestibulocochlear nerve structural organization
nerve development
branchiomotor neuron axon guidance
gonadotrophin-releasing hormone neuronal migration to the hypothalamus
retinal ganglion cell axon guidance
cellular response to hepatocyte growth factor stimulus
endothelial cell chemotaxis
cellular response to vascular endothelial growth factor stimulus
ventral trunk neural crest cell migration
VEGF-activated neuropilin signaling pathway
otic placode formation
negative regulation of neuron apoptotic process
platelet-derived growth factor receptor signaling pathway
vascular endothelial growth factor receptor signaling pathway
hepatocyte growth factor receptor signaling pathway
positive regulation of axon extension involved in axon guidance
negative regulation of axon extension involved in axon guidance
artery morphogenesis
axon extension involved in axon guidance
positive regulation of peptidyl-tyrosine phosphorylation
positive chemotaxis
positive regulation of cytokine activity
axonogenesis involved in innervation
regulation of vesicle-mediated transport
dichotomous subdivision of terminal units involved in salivary gland branching
angiogenesis involved in coronary vascular morphogenesis
coronary artery morphogenesis
retina vasculature morphogenesis in camera-type eye
renal artery morphogenesis
sympathetic ganglion development
trigeminal ganglion development
positive regulation of ERK1 and ERK2 cascade
semaphorin-plexin signaling pathway
commissural neuron axon guidance
regulation of retinal ganglion cell axon guidance
endothelial tip cell fate specification
sensory neuron axon guidance
sympathetic neuron projection extension
sympathetic neuron projection guidance
neural crest cell migration involved in autonomic nervous system development
toxin transport
semaphorin-plexin signaling pathway involved in neuron projection guidance
semaphorin-plexin signaling pathway involved in axon guidance
positive regulation of retinal ganglion cell axon guidance
VEGF-activated neuropilin signaling pathway involved in axon guidance
protein localization to early endosome
facioacoustic ganglion development
dorsal root ganglion morphogenesis
negative regulation of extrinsic apoptotic signaling pathway
vascular endothelial growth factor-activated receptor activity
protein binding
heparin binding
coreceptor activity
semaphorin receptor activity
growth factor binding
cytokine binding
vascular endothelial growth factor binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0