Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ACOX1 SIRNA Assays ›
          Gene Symbol:
          ACOX1
          Gene Name:
          acyl-CoA oxidase 1
          Gene Aliases:
          ACOX, PALMCOX, SCOX
          Chromosome Location:
          Chr.17: 75941507 - 75979434 on Build GRCh38
          UniGene ID:
          Hs.464137
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 107304

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ACOX1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001185039.1 NP_001171968.1 13, 14 2478
          NM_004035.6 NP_004026.2 13, 14 2304
          NM_007292.5 NP_009223.2 13, 14 2304
          XM_011524868.2 XP_011523170.1 14, 15 2463
          XM_011524869.2 XP_011523171.1 12, 13 2141
          GenBank mRNA AK291793.1 - 13, 14 1998
          AK292965.1 - 13, 14 2003
          AK296409.1 - 13, 14 2118
          AK300447.1 - 14, 15 2159
          AK308218.1 - 12, 13 1743
          BC008767.2 - 13, 14 2233 3051501
          BC010425.1 - 13, 14 2022 3357409
          BX537380.1 - 13, 14 1963
          U07866.1 - 13, 14 1943
          X71440.1 - 13, 14 1956

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          oxidoreductase

          Gene Ontology Categories:

          Function(s) Process(es)

          very long-chain fatty acid metabolic process
          generation of precursor metabolites and energy
          lipid metabolic process
          prostaglandin metabolic process
          spermatogenesis
          fatty acid oxidation
          fatty acid beta-oxidation using acyl-CoA dehydrogenase
          fatty acid beta-oxidation using acyl-CoA oxidase
          alpha-linolenic acid metabolic process
          fatty-acyl-CoA binding
          acyl-CoA dehydrogenase activity
          acyl-CoA oxidase activity
          receptor binding
          electron carrier activity
          PDZ domain binding
          protein N-terminus binding
          flavin adenine dinucleotide binding
          oxidoreductase activity, acting on the CH-CH group of donors, with a flavin as acceptor

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0