Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FASN SIRNA Assays ›
          Gene Symbol:
          FASN
          Gene Name:
          fatty acid synthase
          Gene Aliases:
          FAS, OA-519, SDR27X1
          Chromosome Location:
          Chr.17: 82078338 - 82098303 on Build GRCh38
          UniGene ID:
          Hs.83190
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 107316

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: FASN
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_004104.4 NP_004095.4 42 7433
          XM_011523538.2 XP_011521840.1 42 7501
          GenBank mRNA AB209988.1 - 42 7428
          AY451392.1 - 41 7316
          BC007267.1 - 16 2575
          BC007909.1 - 11 1848
          BC014631.2 - 18 3422
          BC014634.2 - 5 844
          BC063242.1 - 42 7433 6172538
          S80437.1 - 7 1187
          U29344.1 - 42 7433

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: NM_007988, AK147214, BC046513, AK147694, X13135, and others.

          Gene Ontology Categories:

          Function(s) Process(es)

          osteoblast differentiation
          acetyl-CoA metabolic process
          fatty acid metabolic process
          fatty acid biosynthetic process
          pantothenate metabolic process
          mammary gland development
          positive regulation of cellular metabolic process
          long-chain fatty-acyl-CoA biosynthetic process
          oxidation-reduction process
          cellular response to interleukin-4
          cell-cell adhesion
          fatty acid synthase activity
          [acyl-carrier-protein] S-acetyltransferase activity
          [acyl-carrier-protein] S-malonyltransferase activity
          3-oxoacyl-[acyl-carrier-protein] synthase activity
          3-oxoacyl-[acyl-carrier-protein] reductase (NADPH) activity
          3-hydroxypalmitoyl-[acyl-carrier-protein] dehydratase activity
          enoyl-[acyl-carrier-protein] reductase (NADPH, B-specific) activity
          oleoyl-[acyl-carrier-protein] hydrolase activity
          protein binding
          drug binding
          myristoyl-[acyl-carrier-protein] hydrolase activity
          palmitoyl-[acyl-carrier-protein] hydrolase activity
          phosphopantetheine binding
          protein homodimerization activity
          poly(A) RNA binding
          enoyl-[acyl-carrier-protein] reductase (NADPH, A-specific) activity
          3-hydroxyoctanoyl-[acyl-carrier-protein] dehydratase activity
          NADPH binding
          cadherin binding involved in cell-cell adhesion
          3-oxo-pimeloyl-[acp] methyl ester reductase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0