Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199509.1 | NP_001186438.1 | 2 | 408 | ||
NM_001199510.1 | NP_001186439.1 | 2 | 408 | |||
NM_004828.3 | NP_004819.2 | 2 | 408 | |||
XM_017011500.1 | XP_016866989.1 | 2 | 578 | |||
GenBank mRNA | AJ010099.1 | - | 2 | 347 | ||
AJ010100.1 | - | 2 | 347 |