Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GATA6 SIRNA Assays ›
Gene Symbol:
GATA6
Gene Name:
GATA binding protein 6
Gene Aliases:
-
Chromosome Location:
Chr.18: 22169437 - 22202528 on Build GRCh38
UniGene ID:
Hs.514746
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
107668
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: GATA6
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_005257.5
NP_005248.2
7
2681
GenBank mRNA
D87811.1
-
6
2221
U66075.1
-
6
2307
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_010258, AK142381, S82462.
Panther Classification:
Molecular Function -
DNA-binding transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
in utero embryonic development
liver development
outflow tract septum morphogenesis
type B pancreatic cell differentiation
pancreatic A cell differentiation
transcription from RNA polymerase II promoter
phospholipid metabolic process
endodermal cell fate determination
blood coagulation
male gonad development
cardiac muscle hypertrophy in response to stress
positive regulation of BMP signaling pathway
negative regulation of transforming growth factor beta1 production
negative regulation of transforming growth factor beta2 production
tube morphogenesis
response to drug
negative regulation of apoptotic process
response to estrogen
cellular protein metabolic process
cell fate commitment
positive regulation of angiogenesis
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
cell development
organ formation
smooth muscle cell differentiation
positive regulation of cardioblast differentiation
cardiac muscle cell differentiation
positive regulation of cardiac muscle cell proliferation
lung saccule development
Clara cell differentiation
Type II pneumocyte differentiation
intestinal epithelial cell differentiation
cardiac vascular smooth muscle cell differentiation
response to growth factor
positive regulation of cell cycle arrest
cellular response to gonadotropin stimulus
cellular response to hypoxia
cellular response to BMP stimulus
RNA polymerase II regulatory region sequence-specific DNA binding
RNA polymerase II core promoter sequence-specific DNA binding
RNA polymerase II transcription factor activity, sequence-specific DNA binding
RNA polymerase II transcription factor binding
RNA polymerase II repressing transcription factor binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
protein binding
transcription factor binding
zinc ion binding
protein kinase binding
transcription regulatory region DNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0