siRNA Details
Gene Symbol: SOX12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006943.3 | NP_008874.2 | 1 | 4628 |

Gene Symbol: SOX12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006943.3 | NP_008874.2 | 1 | 4628 |