Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TFEB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167827.2 | NP_001161299.2 | 9 | 1847 | ||
NM_001271943.1 | NP_001258872.1 | 8 | 1656 | |||
NM_001271944.1 | NP_001258873.1 | 9 | 1911 | |||
NM_001271945.1 | NP_001258874.1 | 9 | 1822 | |||
NM_007162.2 | NP_009093.1 | 10 | 2013 | |||
XM_005249411.1 | XP_005249468.1 | 9 | 1946 | |||
XM_005249412.1 | XP_005249469.1 | 9 | 1838 | |||
XM_006715212.3 | XP_006715275.1 | 9 | 2065 | |||
XM_006715213.2 | XP_006715276.1 | 9 | 1889 | |||
XM_011514915.1 | XP_011513217.1 | 9 | 2068 | |||
XM_011514916.2 | XP_011513218.1 | 10 | 1858 | |||
GenBank mRNA | AJ608786.1 | - | 9 | 1847 | ||
BC006225.2 | - | 7 | 1285 | |||
BC032448.1 | - | 10 | 2013 | 5180066 |