Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDZRN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164595.1 | NP_001158067.1 | 5 | 1156 | ||
NM_013377.3 | NP_037509.3 | 3 | 580 | |||
GenBank mRNA | AK091096.1 | - | 3 | 547 | ||
AK131554.1 | - | 5 | 756 | |||
AK294431.1 | - | 3 | 636 | |||
AK294535.1 | - | 5 | 619 | |||
BC093744.1 | - | 3 | 429 | 7939589 | ||
BC093746.2 | - | 3 | 429 | 7939591 | ||
BX640695.1 | - | 1 | 309 |