Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRP10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005267510.1 | XP_005267567.1 | 7 | 3293 | ||
GenBank mRNA | AF131760.1 | - | 3 | 1506 | ||
AF172816.1 | - | 3 | 2174 | |||
AF177336.1 | - | 3 | 1618 | |||
AK291030.1 | - | 7 | 2723 | |||
AL080164.1 | - | 3 | 1329 | |||
AL110149.1 | - | 1 | 674 | |||
AY358399.1 | - | 7 | 3123 | |||
BC018816.2 | - | 3 | 1688 | |||
BC080648.1 | - | 3 | 1224 | |||
BX161500.1 | - | 7 | 3051 | |||
CD677124.1 | - | 120 |