Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NCOA6 SIRNA Assays ›
          Gene Symbol:
          NCOA6
          Gene Name:
          nuclear receptor coactivator 6
          Gene Aliases:
          AIB3, ASC2, NRC, PRIP, RAP250, TRBP
          Chromosome Location:
          Chr.20: 34714774 - 34825649 on Build GRCh38
          UniGene ID:
          Hs.368971
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 108476

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NCOA6
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001242539.2 NP_001229468.1 12 3506
          NM_001318240.1 NP_001305169.1 16 9007
          NM_014071.4 NP_054790.2 15 6777
          XM_011528721.2 XP_011527023.1 15 6722
          XM_011528726.2 XP_011527028.1 15 6689
          XM_011528727.2 XP_011527029.1 15 6686
          XM_017027739.1 XP_016883228.1 16 6779
          XM_017027740.1 XP_016883229.1 17 6898
          XM_017027741.1 XP_016883230.1 16 6776
          XM_017027742.1 XP_016883231.1 16 6764
          XM_017027743.1 XP_016883232.1 15 6630
          XM_017027744.1 XP_016883233.1 15 6668
          XM_017027745.1 XP_016883234.1 15 6635
          XM_017027747.1 XP_016883236.1 13 6156
          XM_017027750.1 XP_016883239.1 15 3800
          XM_017027751.1 XP_016883240.1 14 3726
          GenBank mRNA AF128458.1 - 14 6638
          AF171667.1 - 14 6580
          AF177388.1 - 14 6596
          AF208227.1 - 16 9004
          AF245115.1 - 13 6495
          AK027040.1 - 5 2734
          BC136272.1 - 14 6586 9020259
          D80003.2 - 14 6573

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          chromatin/chromatin-binding, or -regulatory protein

          Gene Ontology Categories:

          Function(s) Process(es)

          ovarian follicle development
          DNA replication
          DNA repair
          DNA recombination
          DNA-templated transcription, initiation
          transcription initiation from RNA polymerase II promoter
          cellular response to DNA damage stimulus
          brain development
          heart development
          response to hormone
          myeloid cell differentiation
          intracellular estrogen receptor signaling pathway
          positive regulation of insulin secretion involved in cellular response to glucose stimulus
          glucocorticoid receptor signaling pathway
          cellular lipid metabolic process
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          chromatin binding
          transcription coactivator activity
          protein binding
          ligand-dependent nuclear receptor binding
          enzyme binding
          protein domain specific binding
          estrogen receptor binding
          ligand-dependent nuclear receptor transcription coactivator activity
          glucocorticoid receptor binding
          protein homodimerization activity
          peroxisome proliferator activated receptor binding
          retinoid X receptor binding
          thyroid hormone receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-6c5pt:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0