Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128160.1 | NP_001121632.1 | 7 | 1263 | ||
NM_001128161.1 | NP_001121633.1 | 8 | 882 | |||
NM_014517.4 | NP_055332.3 | 7 | 1263 | |||
GenBank mRNA | AF198487.1 | - | 7 | 907 | ||
AK023274.1 | - | 7 | 892 | |||
AK023278.1 | - | 2 | 173 | |||
AK293100.1 | - | 8 | 882 | |||
BC047235.1 | - | 7 | 1262 | 5262788 |