Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MTHFD1L SIRNA Assays ›
Gene Symbol:
MTHFD1L
Gene Name:
methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 1-like
Gene Aliases:
FTHFSDC1, MTC1THFS, dJ292B18.2
Chromosome Location:
Chr.6: 150865679 - 151101887 on Build GRCh38
UniGene ID:
Hs.591343
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
108716
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MTHFD1L
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001242767.1
NP_001229696.1
16
1823
NM_001242768.1
NP_001229697.1
16
1507
NM_015440.4
NP_056255.2
16
1820
XM_005266907.4
XP_005266964.1
16
1761
XM_005266910.4
XP_005266967.1
16
1764
XM_005266911.4
XP_005266968.1
16
1764
XM_011535729.2
XP_011534031.1
16
1764
XM_011535730.2
XP_011534032.1
16
1507
XM_011535731.2
XP_011534033.1
16
1479
XM_011535732.2
XP_011534034.1
16
1561
XM_011535733.2
XP_011534035.1
16
2007
XM_011535734.2
XP_011534036.1
16
1600
XM_011535737.2
XP_011534039.1
16
1764
XM_011535738.2
XP_011534040.1
16
1764
XM_017010702.1
XP_016866191.1
16
1763
XM_017010703.1
XP_016866192.1
16
1763
XM_017010704.1
XP_016866193.1
16
1479
XM_017010705.1
XP_016866194.1
16
2004
XM_017010706.1
XP_016866195.1
16
2007
XM_017010707.1
XP_016866196.1
16
2004
XM_017010708.1
XP_016866197.1
16
1765
GenBank mRNA
AB127387.1
-
16
1676
AK127089.1
-
15
2299
AK315378.1
-
15
1616
AL117452.1
-
16
1495
AY374130.1
-
16
1783
BC008629.1
-
2
89
BC017477.1
-
11
1067
BC110319.1
-
15
1442
BC136685.1
-
16
1713
9020675
BC144368.1
-
16
1713
9052889
BC144369.1
-
16
1710
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
neural tube closure
one-carbon metabolic process
folic acid-containing compound metabolic process
folic acid-containing compound biosynthetic process
formate metabolic process
tetrahydrofolate interconversion
tetrahydrofolate metabolic process
folic acid metabolic process
embryonic neurocranium morphogenesis
embryonic viscerocranium morphogenesis
oxidation-reduction process
formate-tetrahydrofolate ligase activity
methenyltetrahydrofolate cyclohydrolase activity
methylenetetrahydrofolate dehydrogenase (NADP+) activity
ATP binding
protein homodimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0