Search
Search

PAXBP1| Gene Symbol: PAXBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013329.3 | NP_037461.2 | 4 | 949 | ||
| NM_016631.3 | NP_057715.2 | 4 | 949 | |||
| NR_027873.1 | 4 | 949 | ||||
| XM_006724066.1 | XP_006724129.1 | 4 | 878 | |||
| XM_011529804.2 | XP_011528106.1 | 4 | 878 | |||
| GenBank mRNA | AF231920.1 | - | 3 | 366 | ||
| AJ279080.1 | - | 4 | 775 | |||
| AJ279081.1 | - | 4 | 775 | |||
| AK309404.1 | - | 4 | 878 | |||
| AY033903.1 | - | 4 | 759 | |||
| AY033904.1 | - | 4 | 759 | |||
| AY033905.1 | - | 4 | 759 | |||
| AY033906.1 | - | 4 | 759 | |||
| BC030539.1 | - | 4 | 894 | |||