Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SLCO1C1 SIRNA Assays ›
Gene Symbol:
SLCO1C1
Gene Name:
solute carrier organic anion transporter family member 1C1
Gene Aliases:
OATP-F, OATP1, OATP14, OATP1C1, OATPF, OATPRP5, SLC21A14
Chromosome Location:
Chr.12: 20690505 - 20753386 on Build GRCh38
UniGene ID:
Hs.47261
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
108945
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SLCO1C1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001145944.1
NP_001139416.1
8, 9
1461
NM_001145945.1
NP_001139417.1
10, 11
1597
NM_001145946.1
NP_001139418.1
11, 12
1744
NM_017435.4
NP_059131.1
10, 11
1757
XM_005253394.2
XP_005253451.1
11, 12
1533
XM_005253396.2
XP_005253453.1
9, 10
6298
XM_011520703.2
XP_011519005.1
11, 12
1533
XM_011520704.2
XP_011519006.1
11, 12
1533
XM_011520709.2
XP_011519011.1
8, 9
1175
XM_011520711.2
XP_011519013.1
10, 11
6353
XM_017019484.1
XP_016874973.1
13, 14
6794
XM_017019485.1
XP_016874974.1
12, 13
6712
XM_017019486.1
XP_016874975.1
10, 11
6452
XM_017019487.1
XP_016874976.1
9, 10
6298
XM_017019488.1
XP_016874977.1
14, 15
6849
XM_017019489.1
XP_016874978.1
15, 16
6949
XM_017019490.1
XP_016874979.1
15, 16
6913
GenBank mRNA
AF205076.1
-
10, 11
1758
AF260704.1
-
10, 11
1453
AK294333.1
-
8, 9
1329
AK296236.1
-
10, 11
1490
AK303713.1
-
11, 12
1620
AK308729.1
-
9, 10
1393
AL834209.1
-
11, 12
1609
BC022461.2
-
10, 11
1486
4801171
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_053441, XM_006237602, AF306546.
This siRNA also targets the following transcripts in Mouse: NM_021471, NM_001177772, AK048926, AK161010, BE865248, and others.
Panther Classification:
Molecular Function -
transporter
Gene Ontology Categories:
Function(s)
Process(es)
bile acid metabolic process
bile acid and bile salt transport
sodium-independent organic anion transport
thyroid hormone transport
bile acid transmembrane transporter activity
sodium-independent organic anion transmembrane transporter activity
thyroid hormone transmembrane transporter activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-qm47m:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0