Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CADPS2 SIRNA Assays ›
          Gene Symbol:
          CADPS2
          Gene Name:
          calcium dependent secretion activator 2
          Gene Aliases:
          CAPS2
          Chromosome Location:
          Chr.7: 122318424 - 122886759 on Build GRCh38
          UniGene ID:
          Hs.649459
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 109027

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CADPS2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001009571.3 NP_001009571.2 6 1556
          NM_001167940.1 NP_001161412.1 6 1556
          NM_017954.10 NP_060424.9 6 1556
          XM_005250695.4 XP_005250752.1 6 1285
          XM_005250696.4 XP_005250753.1 6 1285
          XM_005250697.4 XP_005250754.1 6 1285
          XM_005250698.4 XP_005250755.1 6 1257
          XM_005250699.4 XP_005250756.1 6 1285
          XM_005250700.4 XP_005250757.1 6 1285
          XM_005250701.4 XP_005250758.1 6 1285
          XM_005250702.4 XP_005250759.1 6 1285
          XM_005250703.4 XP_005250760.1 6 1285
          XM_005250704.4 XP_005250761.1 6 1285
          XM_005250705.4 XP_005250762.1 6 1285
          XM_005250706.4 XP_005250763.1 6 1285
          XM_005250707.4 XP_005250764.1 6 1285
          XM_005250708.3 XP_005250765.1 6 1285
          XM_011516695.2 XP_011514997.1 6 1257
          XM_011516696.2 XP_011514998.1 6 1285
          XM_011516697.2 XP_011514999.1 6 1285
          XM_017012794.1 XP_016868283.1 6 1285
          XM_017012795.1 XP_016868284.1 6 1285
          XM_017012796.1 XP_016868285.1 7 900
          XM_017012797.1 XP_016868286.1 7 966
          XM_017012798.1 XP_016868287.1 6 1436
          XM_017012799.1 XP_016868288.1 7 1337
          XM_017012800.1 XP_016868289.1 3 293
          GenBank mRNA AB046811.1 - 4 425
          AF401638.1 - 6 1225
          AK025672.1 - 2 79
          AK126518.1 - 7 1158
          AL833058.1 - 3 351
          AY264289.1 - 6 1160
          BC136601.1 - 6 1255 9020591
          BC144278.1 - 6 1255 9052798
          BC144279.1 - 6 1255 9052799
          KJ534798.1 - 6 1134
          KJ534800.1 - 6 1134
          KJ534801.1 - 6 1134
          KJ534802.1 - 6 1134
          KJ534803.1 - 6 1134
          KJ534804.1 - 6 1134
          KJ534805.1 - 6 1134

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          membrane trafficking regulatory protein

          Gene Ontology Categories:

          Function(s) Process(es)

          cellular response to starvation
          protein transport
          synaptic vesicle priming
          positive regulation of exocytosis
          dense core granule exocytosis
          lipid binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0