Search
Search

SLC35C1| Gene Symbol: SLC35C1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145265.1 | NP_001138737.1 | 3 | 2079 | ||
| NM_001145266.1 | NP_001138738.1 | 3 | 1982 | |||
| NM_018389.4 | NP_060859.4 | 2 | 2353 | |||
| XM_011520202.2 | XP_011518504.1 | 2 | 1240 | |||
| GenBank mRNA | AK000527.1 | - | 1 | 898 | ||
| AK074929.1 | - | 2 | 2354 | |||
| BC001427.2 | - | 2 | 1658 | 3139599 | ||