siRNA Details
Gene Symbol: GABRB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000813.2 | NP_000804.1 | 7, 8 | 905 | ||
NM_021911.2 | NP_068711.1 | 7, 8 | 905 | |||
GenBank mRNA | A28108.1 | - | 7, 8 | 911 | ||
AK289730.1 | - | 6, 7 | 709 | |||
AK289815.1 | - | 6, 7 | 767 | |||
AK294437.1 | - | 4, 5 | 429 | |||
BC099705.1 | - | 7, 8 | 1070 | 40006771 | ||
BC099719.1 | - | 7, 8 | 1070 | 40006779 | ||
BC105639.1 | - | 7, 8 | 1070 | 40006778 | ||
GU086163.1 | - | 6, 7 | 687 | |||
GU086164.1 | - | 5, 6 | 631 | |||
S67368.1 | - | 7, 8 | 905 | |||
S77553.1 | - | 6, 7 | 687 | |||
S77554.1 | - | 6, 7 | 687 |

More Information
Panther Classification:
Molecular Function -






