Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Instrument Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
Our Instagram
Our Facebook
Blog Behind the Bench
Customer Experience Center (CEC)
Ecommerce Exclusives
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RFWD2 SIRNA Assays ›
Gene Symbol:
RFWD2
Gene Name:
ring finger and WD repeat domain 2
Gene Aliases:
COP1, RNF200
Chromosome Location:
Chr.1: 175944826 - 176207279 on Build GRCh38
UniGene ID:
Hs.523744
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
109471
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RFWD2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001001740.3
NP_001001740.1
10
1381
NM_001286644.1
NP_001273573.1
9
1256
NM_022457.6
NP_071902.2
11
1453
XM_005245447.2
XP_005245504.1
11
1476
XM_005245448.2
XP_005245505.1
10
1428
XM_006711487.2
XP_006711550.1
10
1419
XM_011509881.1
XP_011508183.1
12
1542
XM_017002059.1
XP_016857548.1
12
1542
XM_017002060.1
XP_016857549.1
12
1530
XM_017002061.1
XP_016857550.1
11
1488
XM_017002062.1
XP_016857551.1
11
1482
XM_017002063.1
XP_016857552.1
11
1476
XM_017002064.1
XP_016857553.1
11
1473
XM_017002065.1
XP_016857554.1
10
1428
XM_017002066.1
XP_016857555.1
10
1419
XM_017002067.1
XP_016857556.1
10
1416
XM_017002068.1
XP_016857557.1
10
1413
XM_017002069.1
XP_016857558.1
6
1056
XM_017002070.1
XP_016857559.1
11
1248
XM_017002071.1
XP_016857560.1
9
637
XM_017002072.1
XP_016857561.1
9
635
XM_017002073.1
XP_016857562.1
8
577
XM_017002074.1
XP_016857563.1
8
575
XM_017002075.1
XP_016857564.1
8
657
XM_017002076.1
XP_016857565.1
9
640
XM_017002077.1
XP_016857566.1
7
432
XM_017002078.1
XP_016857567.1
9
705
XM_017002079.1
XP_016857568.1
9
1120
XM_017002080.1
XP_016857569.1
5
359
GenBank mRNA
AF508940.1
-
11
1187
AF527539.1
-
11
1450
AK001278.1
-
9
692
AK314750.1
-
11
1385
AY509921.1
-
10
1115
BC039723.1
-
9
703
BC094728.1
-
11
1441
5271629
BX537932.1
-
1
15
KJ534928.1
-
11
1187
KJ534929.1
-
8
992
KJ534930.1
-
10
1127
KJ534931.1
-
10
1118
KJ534932.1
-
11
1175
KJ534933.1
-
5
686
KJ535061.1
-
8
892
KJ535076.1
-
10
1089
Back To Top
More Information
Panther Classification:
Molecular Function -
ubiquitin-protein ligase
Gene Ontology Categories:
Function(s)
Process(es)
response to ionizing radiation
protein ubiquitination
positive regulation of proteasomal ubiquitin-dependent protein catabolic process
ubiquitin-protein transferase activity
protein binding
zinc ion binding
ligase activity
ubiquitin protein ligase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-qm6d9:80/100.66.79.83:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline