Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RFWD2 SIRNA Assays ›
Gene Symbol:
RFWD2
Gene Name:
ring finger and WD repeat domain 2
Gene Aliases:
COP1, RNF200
Chromosome Location:
Chr.1: 175944826 - 176207279 on Build GRCh38
UniGene ID:
Hs.523744
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
109472
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RFWD2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001001740.3
NP_001001740.1
15
1991
NM_001286644.1
NP_001273573.1
14
1866
NM_022457.6
NP_071902.2
16
2063
XM_005245447.2
XP_005245504.1
16
2086
XM_005245448.2
XP_005245505.1
15
2038
XM_006711487.2
XP_006711550.1
15
2029
XM_011509881.1
XP_011508183.1
17
2152
XM_017002059.1
XP_016857548.1
17
2152
XM_017002060.1
XP_016857549.1
17
2140
XM_017002061.1
XP_016857550.1
16
2098
XM_017002062.1
XP_016857551.1
16
2092
XM_017002063.1
XP_016857552.1
16
2086
XM_017002064.1
XP_016857553.1
16
2083
XM_017002065.1
XP_016857554.1
15
2038
XM_017002066.1
XP_016857555.1
15
2029
XM_017002067.1
XP_016857556.1
15
2026
XM_017002068.1
XP_016857557.1
15
2023
XM_017002069.1
XP_016857558.1
11
1666
XM_017002070.1
XP_016857559.1
16
1858
XM_017002071.1
XP_016857560.1
14
1247
XM_017002072.1
XP_016857561.1
14
1245
XM_017002073.1
XP_016857562.1
13
1187
XM_017002074.1
XP_016857563.1
13
1185
XM_017002075.1
XP_016857564.1
13
1267
XM_017002076.1
XP_016857565.1
14
1250
XM_017002077.1
XP_016857566.1
12
1042
XM_017002078.1
XP_016857567.1
14
1315
XM_017002079.1
XP_016857568.1
14
1730
XM_017002080.1
XP_016857569.1
10
969
GenBank mRNA
AF508940.1
-
16
1797
AF527539.1
-
16
2060
AK001278.1
-
14
1301
AK314750.1
-
16
1995
AY509921.1
-
15
1725
BC039723.1
-
14
1313
BC094728.1
-
16
2051
5271629
BX537932.1
-
5
461
KJ534928.1
-
16
1797
KJ534929.1
-
13
1602
KJ534930.1
-
15
1737
KJ534931.1
-
15
1728
KJ534932.1
-
16
1785
KJ534933.1
-
10
1296
KJ535061.1
-
13
1502
KJ535076.1
-
15
1699
Back To Top
More Information
Panther Classification:
Molecular Function -
ubiquitin-protein ligase
Gene Ontology Categories:
Function(s)
Process(es)
response to ionizing radiation
protein ubiquitination
positive regulation of proteasomal ubiquitin-dependent protein catabolic process
ubiquitin-protein transferase activity
protein binding
zinc ion binding
ligase activity
ubiquitin protein ligase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-5gxp2:80/100.66.75.127:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline