Search
Search

OGFRL1| Gene Symbol: OGFRL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001324266.1 | NP_001311195.1 | 5 | 1008 | ||
| NM_024576.4 | NP_078852.3 | 5 | 1005 | |||
| XM_011536117.2 | XP_011534419.1 | 5 | 941 | |||
| XM_011536118.2 | XP_011534420.1 | 5 | 790 | |||
| XM_017011303.1 | XP_016866792.1 | 5 | 570 | |||
| XM_017011304.1 | XP_016866793.1 | 5 | 567 | |||
| XM_017011305.1 | XP_016866794.1 | 5 | 463 | |||
| XM_017011306.1 | XP_016866795.1 | 5 | 1493 | |||
| GenBank mRNA | AK024732.1 | - | 2 | 68 | ||
| BC018688.1 | - | 5 | 640 | 4717736 | ||
| BC110999.1 | - | 5 | 644 | 5585901 | ||
| BX420584.2 | - | 5 | 674 | |||