Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZEB1 SIRNA Assays ›
Gene Symbol:
ZEB1
Gene Name:
zinc finger E-box binding homeobox 1
Gene Aliases:
AREB6, BZP, DELTAEF1, FECD6, NIL2A, PPCD3, TCF8, ZFHEP, ZFHX1A
Chromosome Location:
Chr.10: 31318495 - 31529814 on Build GRCh38
UniGene ID:
Hs.124503
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
109651
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZEB1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001128128.2
NP_001121600.1
7
1264
NM_001174093.1
NP_001167564.1
6
924
NM_001174094.1
NP_001167565.1
7
1261
NM_001174095.1
NP_001167566.1
6
783
NM_001174096.1
NP_001167567.1
7
987
NM_001323638.1
NP_001310567.1
11
1634
NM_001323641.1
NP_001310570.1
7
1214
NM_001323642.1
NP_001310571.1
9
1516
NM_001323643.1
NP_001310572.1
9
1294
NM_001323644.1
NP_001310573.1
6
1013
NM_001323645.1
NP_001310574.1
8
1357
NM_001323646.1
NP_001310575.1
10
1595
NM_001323647.1
NP_001310576.1
8
1127
NM_001323648.1
NP_001310577.1
7
1067
NM_001323649.1
NP_001310578.1
8
1130
NM_001323650.1
NP_001310579.1
10
1362
NM_001323651.1
NP_001310580.1
8
1030
NM_001323652.1
NP_001310581.1
9
1179
NM_001323653.1
NP_001310582.1
9
1286
NM_001323654.1
NP_001310583.1
9
1173
NM_001323655.1
NP_001310584.1
9
1283
NM_001323656.1
NP_001310585.1
7
958
NM_001323657.1
NP_001310586.1
7
961
NM_001323658.1
NP_001310587.1
7
800
NM_001323659.1
NP_001310588.1
7
1018
NM_001323660.1
NP_001310589.1
7
1015
NM_001323661.1
NP_001310590.1
8
1161
NM_001323662.1
NP_001310591.1
9
1563
NM_001323663.1
NP_001310592.1
10
1502
NM_001323664.1
NP_001310593.1
9
1486
NM_001323665.1
NP_001310594.1
8
1404
NM_001323666.1
NP_001310595.1
7
1344
NM_001323671.1
NP_001310600.1
8
1240
NM_001323672.1
NP_001310601.1
9
1292
NM_001323673.1
NP_001310602.1
8
1237
NM_001323674.1
NP_001310603.1
5
762
NM_001323675.1
NP_001310604.1
5
872
NM_001323676.1
NP_001310605.1
7
1097
NM_001323677.1
NP_001310606.1
7
1094
NM_001323678.1
NP_001310607.1
5
1039
NM_030751.5
NP_110378.3
7
984
XM_006717498.2
XP_006717561.1
10
1422
XM_011519643.2
XP_011517945.1
10
1725
XM_017016597.1
XP_016872086.1
10
1324
XM_017016598.1
XP_016872087.1
10
4454
XM_017016599.1
XP_016872088.1
9
4292
XM_017016600.1
XP_016872089.1
11
4533
XM_017016601.1
XP_016872090.1
10
4451
XM_017016602.1
XP_016872091.1
11
4530
XM_017016603.1
XP_016872092.1
10
4368
GenBank mRNA
AK091478.1
-
6
884
AK294775.1
-
7
1179
AK296244.1
-
7
907
AK299541.1
-
4
521
AK299666.1
-
7
1018
AK300830.1
-
6
731
AK307510.1
-
8
990
AK311137.1
-
5
723
AK314683.1
-
7
945
AL831979.1
-
9
1242
BC009572.1
-
7
940
BC065705.1
-
7
945
BC107781.1
-
7
940
BC112392.1
-
7
940
40036600
BX647794.1
-
11
1635
U12170.1
-
7
945
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: U51584, U51583.
This siRNA also targets the following transcripts in Mouse: NM_011546, AK141437, BC139769, BC090987, AK140038, and others.
Panther Classification:
Molecular Function -
C2H2 zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
immune response
pattern specification process
central nervous system development
cell proliferation
negative regulation of cell proliferation
regulation of mesenchymal cell proliferation
regulation of transforming growth factor beta receptor signaling pathway
negative regulation of epithelial cell differentiation
regulation of T cell differentiation in thymus
positive regulation of neuron differentiation
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
embryonic camera-type eye morphogenesis
embryonic skeletal system morphogenesis
semicircular canal morphogenesis
regulation of smooth muscle cell differentiation
cartilage development
cellular response to amino acid stimulus
cochlea morphogenesis
transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
transcription coactivator activity
transcription corepressor activity
protein binding
transcription factor binding
zinc ion binding
E-box binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0