Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCRL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031282.2 | NP_112572.1 | 12 | 1837 | ||
XM_011510034.1 | XP_011508336.1 | 12 | 1813 | |||
GenBank mRNA | AF329492.1 | - | 9 | 1660 | ||
AF343659.1 | - | 12 | 1837 | |||
AF343660.1 | - | 12 | 1837 | |||
AF343661.1 | - | 12 | 1837 | |||
BC125173.1 | - | 12 | 1795 | 40121786 | ||
BC125174.1 | - | 12 | 1795 | 40121790 |