Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF4A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001967.3 | NP_001958.2 | 8 | 872 | ||
GenBank mRNA | AB209021.1 | - | 3 | 2561 | ||
AF208852.1 | - | 7 | 710 | |||
AK130087.1 | - | 8 | 867 | |||
AK225646.1 | - | 8 | 867 | |||
AK296279.1 | - | 4 | 406 | |||
AL117412.1 | - | 7 | 732 | |||
AL137681.1 | - | 3 | 1437 | |||
AL556981.3 | - | 7 | 732 | |||
BC012547.1 | - | 8 | 874 | 4079371 | ||
BC013708.1 | - | 8 | 852 | 4341420 | ||
BC015842.1 | - | 8 | 871 | 4337581 | ||
BC016295.2 | - | 2 | 150 | |||
BC048105.1 | - | 8 | 870 | 4820774 | ||
BT009860.1 | - | 8 | 833 | |||
D30655.1 | - | 8 | 848 | |||
S79942.1 | - | 8 | 848 |