Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SLC36A1 SIRNA Assays ›
Gene Symbol:
SLC36A1
Gene Name:
solute carrier family 36 member 1
Gene Aliases:
Dct1, LYAAT1, PAT1, TRAMD3
Chromosome Location:
Chr.5: 151411344 - 151556318 on Build GRCh38
UniGene ID:
Hs.269004
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
109925
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SLC36A1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001308150.1
NP_001295079.1
6, 7
735
NM_001308151.1
NP_001295080.1
6, 7
705
NM_078483.3
NP_510968.2
6, 7
735
XM_005268386.1
XP_005268443.1
6, 7
705
XM_006714759.3
XP_006714822.1
6, 7
784
XM_011537580.2
XP_011535882.1
6, 7
1425
XM_011537581.1
XP_011535883.1
7, 8
853
XM_011537583.2
XP_011535885.1
7, 8
5290
XM_011537584.2
XP_011535886.1
6, 7
944
XM_011537585.1
XP_011535887.1
7, 8
800
XM_011537586.2
XP_011535888.1
6, 7
1548
XM_011537587.2
XP_011535889.1
6, 7
804
XM_011537588.2
XP_011535890.1
6, 7
648
XM_011537589.2
XP_011535891.1
6, 7
1556
XM_011537590.1
XP_011535892.1
6, 7
563
XM_011537591.1
XP_011535893.1
6, 7
538
XM_011537592.2
XP_011535894.1
6, 7
542
XM_011537593.2
XP_011535895.1
7, 8
798
XM_011537594.1
XP_011535896.1
8, 9
912
XM_011537595.2
XP_011535897.2
6, 7
1431
XM_011537596.2
XP_011535898.2
5, 6
517
XM_017009216.1
XP_016864705.1
6, 7
631
XM_017009217.1
XP_016864706.1
6, 7
735
XM_017009218.1
XP_016864707.1
5, 6
517
XM_017009219.1
XP_016864708.1
6, 7
785
XM_017009220.1
XP_016864709.1
6, 7
705
GenBank mRNA
AF516142.1
-
5, 6
527
AK057340.1
-
6, 7
705
AK309323.1
-
6, 7
707
AY162213.1
-
6, 7
640
AY227111.1
-
7, 8
655
AY227112.1
-
23
AY227113.1
-
6, 7
611
BC136437.1
-
6, 7
606
9020426
BC136438.1
-
6, 7
606
9020427
BX537963.1
-
6, 7
732
GU296670.1
-
5, 6
512
Back To Top
More Information
Panther Classification:
Molecular Function -
amino acid transporter
Gene Ontology Categories:
Function(s)
Process(es)
ion transport
amino acid transport
L-alanine transport
glycine transport
proline transport
proline transmembrane transport
hydrogen ion transmembrane transport
hydrogen:amino acid symporter activity
hydrogen ion transmembrane transporter activity
amino acid transmembrane transporter activity
L-alanine transmembrane transporter activity
glycine transmembrane transporter activity
L-proline transmembrane transporter activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-zvggx:80/10.239.163.62:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0