Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other BRD8 SIRNA Assays ›
          Gene Symbol:
          BRD8
          Gene Name:
          bromodomain containing 8
          Gene Aliases:
          SMAP, SMAP2, p120
          Chromosome Location:
          Chr.5: 138139766 - 138178669 on Build GRCh38
          UniGene ID:
          Hs.519337
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 110031

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: BRD8
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001164326.1 NP_001157798.1 5, 6 411
          NM_001300961.1 NP_001287890.1 5, 6 416
          NM_001300962.1 NP_001287891.1 4, 5 341
          NM_001300966.1 NP_001287895.1 5, 6 411
          NM_006696.3 NP_006687.3 5, 6 411
          NM_139199.1 NP_631938.1 5, 6 411
          XM_005271855.2 XP_005271912.1 5, 6 402
          XM_005271856.2 XP_005271913.1 5, 6 402
          XM_005271857.2 XP_005271914.1 5, 6 402
          XM_005271859.3 XP_005271916.1 5, 6 402
          XM_005271860.3 XP_005271917.1 5, 6 402
          XM_005271861.3 XP_005271918.1 5, 6 402
          XM_011543109.1 XP_011541411.1 5, 6 402
          XM_011543110.1 XP_011541412.1 5, 6 388
          XM_017008969.1 XP_016864458.1 5, 6 402
          XM_017008970.1 XP_016864459.1 5, 6 402
          XM_017008971.1 XP_016864460.1 5, 6 402
          XM_017008972.1 XP_016864461.1 5, 6 402
          XM_017008973.1 XP_016864462.1 5, 6 402
          XM_017008974.1 XP_016864463.1 5, 6 402
          XM_017008975.1 XP_016864464.1 5, 6 402
          XM_017008976.1 XP_016864465.1 5, 6 402
          GenBank mRNA AB209079.1 - 5, 6 357
          AF016270.1 - 4, 5 343
          AK225928.1 - 5, 6 374
          AK289951.1 - 5, 6 371
          AK293621.1 - 4, 5 301
          AK297756.1 - 5, 6 376
          AK310380.1 - 3, 4 250
          AK312333.1 - 5, 6 371
          AL136823.1 - 5, 6 384
          AL704983.1 - 5, 6 411
          BC008039.2 - 5, 6 353 3009915
          BC008076.2 - 5, 6 353 3009915
          BU187982.1 - 6, 7 555
          X87613.1 - 4, 5 341
          X94234.1 - 4, 5 341

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          signal transduction
          cell surface receptor signaling pathway
          intracellular receptor signaling pathway
          regulation of growth
          histone H4 acetylation
          histone H2A acetylation
          transcription factor activity, sequence-specific DNA binding
          thyroid hormone receptor activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0