Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177478.1 | NP_001170949.1 | 3 | 817 | ||
NM_001177479.1 | NP_001170950.1 | 4 | 964 | |||
NM_144657.4 | NP_653258.2 | 3 | 855 | |||
XM_006724619.1 | XP_006724682.1 | 2 | 708 | |||
XM_011530863.2 | XP_011529165.1 | 4 | 1037 | |||
XM_011530864.1 | XP_011529166.1 | 4 | 944 | |||
XM_017029274.1 | XP_016884763.1 | 3 | 835 | |||
XM_017029275.1 | XP_016884764.1 | 3 | 835 | |||
GenBank mRNA | AK055240.1 | - | 3 | 791 | ||
AK290050.1 | - | 3 | 829 | |||
BC101720.1 | - | 3 | 767 | 8069226 | ||
BC101722.1 | - | 3 | 767 | 8069228 | ||
BC143522.1 | - | 4 | 876 | 9052031 | ||
BX538112.1 | - | 3 | 827 | |||
BX649166.1 | - | 3 | 830 |