siRNA Details
Gene Symbol: CAMKK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032294.2 | NP_115670.1 | 6 | 750 | ||
NM_172206.1 | NP_757343.1 | 6 | 696 | |||
NM_172207.2 | NP_757344.2 | 6 | 750 | |||
XM_005256817.1 | XP_005256874.1 | 6 | 750 | |||
XM_006721588.2 | XP_006721651.1 | 6 | 1310 | |||
XM_011524031.2 | XP_011522333.2 | 6 | 869 | |||
XM_011524032.2 | XP_011522334.1 | 6 | 947 | |||
XM_011524033.1 | XP_011522335.1 | 5 | 648 | |||
XM_017025219.1 | XP_016880708.1 | 6 | 859 | |||
XM_017025220.1 | XP_016880709.1 | 6 | 1310 | |||
XM_017025222.1 | XP_016880711.1 | 6 | 927 | |||
GenBank mRNA | AF425232.1 | - | 6 | 703 | ||
AF425301.1 | - | 6 | 696 | |||
AK128601.1 | - | 6 | 1310 | |||
AL136576.1 | - | 6 | 750 | |||
BC031647.1 | - | 6 | 684 | 5175128 | ||
BC043487.1 | - | 6 | 726 | 5751570 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
