Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC5A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145913.3 | NP_666018.3 | 15 | 2746 | ||
GenBank mRNA | AF536216.1 | - | 15 | 2721 |