siRNA Details
Gene Symbol: CDYL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152342.3 | NP_689555.2 | 2 | 468 | ||
XM_011522866.1 | XP_011521168.1 | 2 | 480 | |||
XM_011522867.2 | XP_011521169.1 | 2 | 1023 | |||
GenBank mRNA | AK096185.1 | - | 2 | 468 | ||
AX781670.1 | - | 2 | 455 | |||
AX781671.1 | - | 2 | 363 | |||
AY273798.1 | - | 2 | 508 | |||
BC069440.1 | - | 2 | 413 | 7262286 | ||
BC100803.1 | - | 2 | 387 | 40000745 | ||
BC100804.1 | - | 2 | 387 | 40000746 | ||
BC100805.1 | - | 2 | 387 | 40000747 | ||
BC100806.1 | - | 2 | 387 | 40000748 |

More Information
Panther Classification:
Molecular Function -










