Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC9B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006714085.2 | XP_006714148.1 | 12, 13 | 2255 | ||
GenBank mRNA | BC047447.1 | - | 12, 13 | 1786 | 5295722 |