Search
Search

SEZ6| Gene Symbol: SEZ6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001098635.1 | NP_001092105.1 | 2, 3 | 926 | ||
| NM_001290202.1 | NP_001277131.1 | 2, 3 | 762 | |||
| NM_178860.4 | NP_849191.3 | 2, 3 | 926 | |||
| XM_011524315.1 | XP_011522617.1 | 2, 3 | 926 | |||
| XM_011524317.2 | XP_011522619.1 | 2, 3 | 926 | |||
| GenBank mRNA | AF502129.1 | - | 2, 3 | 1014 | ||
| AF502130.1 | - | 2, 3 | 1014 | |||
| AK091522.1 | - | 2, 3 | 762 | |||
| AK096473.1 | - | 1, 2 | 230 | |||
| AK125377.1 | - | 2, 3 | 888 | |||
| AK223620.1 | - | 2, 3 | 888 | |||
| AL834405.1 | - | 1, 2 | 671 | |||
| AY038048.1 | - | 2, 3 | 917 | |||