Search
Search

SEZ6| Gene Symbol: SEZ6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001098635.1 | NP_001092105.1 | 17 | 3562 | ||
| NM_001290202.1 | NP_001277131.1 | 17 | 3414 | |||
| NM_178860.4 | NP_849191.3 | 17 | 3578 | |||
| XM_011524315.1 | XP_011522617.1 | 16 | 3459 | |||
| XM_011524317.2 | XP_011522619.1 | 16 | 3475 | |||
| GenBank mRNA | AF502129.1 | - | 17 | 3666 | ||
| AK054913.1 | - | 8 | 1439 | |||
| AK055383.1 | - | 11 | 1850 | |||
| AK091522.1 | - | 17 | 3414 | |||
| AK096473.1 | - | 16 | 2811 | |||
| AL834405.1 | - | 16 | 3307 | |||
| AY038048.1 | - | 17 | 3569 | |||