Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000367.3 | NP_000358.1 | 7, 8 | 765 | ||
XM_017011241.1 | XP_016866730.1 | 8, 9 | 1059 | |||
GenBank mRNA | AF021876.1 | - | 6, 7 | 588 | ||
AF021877.1 | - | 6, 7 | 588 | |||
BC005339.1 | - | 7, 8 | 675 | 3829338 | ||
BC009596.1 | - | 7, 8 | 674 | 4133029 | ||
S62904.1 | - | 6, 7 | 653 | |||
U12387.1 | - | 7, 8 | 664 |